Merck
  • Butalbital in the treatment of headache: history, pharmacology, and efficacy.
Butalbital in the treatment of headache: history, pharmacology, and efficacy.
Headache (2002-03-21)
S D Silberstein, D C McCrory
ABSTRACT
Analgesics containing butalbital compounded with aspirin, acetaminophen, and/or caffeine are widely used for the treatment of migraine and tension-type headache. The butalbital-containing compounds are efficacious in placebo-controlled trials among patients with episodic tension-type headaches. Despite their frequent clinical use for migraine, they have not been studied in placebo-controlled trials among patients with migraine. Barbiturates can produce intoxication, hangover, tolerance, dependence, and toxicity. Butalbital can result in intoxication that is clinically indistinguishable from that produced by alcohol. Butalbital-containing analgesics can produce drug-induced headache in addition to tolerance and dependence. Higher doses can produce withdrawal syndromes after discontinuation. Butalbital-containing analgesics may be effective as backup medications or when other medications are ineffective or cannot be used. Because of concerns about overuse, medication-overuse headache, and withdrawal, their use should be limited and carefully monitored.
Product Description
Supelco
Butalbital solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®