Merck
  • Benzannulation for the regiodefined synthesis of 2-alkyl/aryl-1-naphthols: total synthesis of arnottin I.
Benzannulation for the regiodefined synthesis of 2-alkyl/aryl-1-naphthols: total synthesis of arnottin I.
The Journal of organic chemistry (2011-03-17)
Dipakranjan Mal, Amit Kumar Jana, Prithiba Mitra, Ketaki Ghosh
ABSTRACT
The annulation of phthalides with α-alkyl/arylacrylates in the presence of LDA/LHMDS is shown to directly give alkyl/aryl-1-naphthols. The method involving a novel dealkoxycarbonylation obviates the regiochemical issues in the synthesis of polysubstituted naphthalenes, and it forms the key step in a three-step total synthesis of arnottin I, a naphthobenzopyranone natural product.
Sigma-Aldrich
Phthalide, 98%