Merck
  • Synthesis of sulfuric macrocycles and a rotaxane through thiol-yne click and dithiol coupling reactions.
Synthesis of sulfuric macrocycles and a rotaxane through thiol-yne click and dithiol coupling reactions.
Organic letters (2010-08-18)
Weidong Zhou, Haiyan Zheng, Yongjun Li, Huibiao Liu, Yuliang Li
ABSTRACT
A macrocycle and a rotaxane were constructed by virtue of the thiol-yne click reaction under the irradiation of light in high yield, which can proceed at ambient temperature and humidity under an air atmosphere. Two disulfide macrocycles were synthesized through a simple dithiol coupling reaction, which exhibited high stability and a weak assembly interaction with a dialkylammonium ion.
Sigma-Aldrich
Toluene-3,4-dithiol, technical grade, 90%
Supelco
Toluene-3,4-dithiol, for spectrophotometric det. of Mo, Sn, W, and also Ag and Re, ≥97.0%