Merck
  • Anti-inflammatory role of extracellular l-arginine through calcium sensing receptor in human renal proximal tubular epithelial (HK-2) cells.
Anti-inflammatory role of extracellular l-arginine through calcium sensing receptor in human renal proximal tubular epithelial (HK-2) cells.
International immunopharmacology (2023-02-25)
Samuel Shin, Eugenia Awuah Boadi, Saloni Shah, Madison Ezell, Peijun Li, Bidhan C Bandyopadhyay
ABSTRACT
Renal tubular epithelial cells are capable of synthesizing interleukins (IL) in response to a variety of proinflammatory cytokines. Moreover, elevated urinary levels of IL have been shown in patients with various forms of nephritic diseases. However, the underlying intracellular signaling mechanism is unclear. Here we show the immunological signaling role of l-Arginine (l-Arg) through Ca2+-sensing receptor (CaSR) in human kidney 2 (HK-2) renal proximal tubular epithelial cells, using Ca2+ imaging and patch clamp techniques and its mechanistic link to the downstream cellular function. Both pharmacological and siRNA inhibitors support the activation CaSR by extracellular l-Arg to induced Ca2+ entry via a Transient receptor potential canonical (TRPC) channel in HK-2 cells mainly through the receptor operated Ca2+ entry (ROCE). Activation of CaSR by l-Arg led to the rise in p-p38/p38 expression suggesting [Ca2+]i as a regulator for p38-signaling pathways. Notably, l-Arg activated CaSR-induced Ca2+ signaling reduced the expressions of key fibrotic, inflammatory, and apoptotic genes, suggesting its nephroprotective role via Ca2+ signaling through CaSR in HK-2 cells. Since we found that the IL-6 expressions were inversely proportional to the increasing concentrations of l-Arg in HK-2 cells, we measured the release of IL-6, which steadily decreased as the concentrations of l-Arg were elevated. Taken together, extracellular l-Arg is a negative regulator for IL-6-induced inflammatory process, through the activation of CaSR and TRPC channel by ROCE pathway and can have a potential to alleviate inflammatory renal diseases.
Sigma-Aldrich
Pyr10, ≥98% (HPLC)
Sigma-Aldrich
2-Aminoethyl diphenylborinate, 97%