Merck
  • In vitro and in vivo antibacterial activities of ME1207, a new oral cephalosporin.
In vitro and in vivo antibacterial activities of ME1207, a new oral cephalosporin.
Antimicrobial agents and chemotherapy (1988-09-01)
A Tamura, R Okamoto, T Yoshida, H Yamamoto, S Kondo, M Inoue, S Mitsuhashi
ABSTRACT
ME1207 (pivaloyloxymethyl ester of ME1206) is a new oral cephalosporin. ME1206 is (6R,7R)-7-[(Z)-2-(2-aminothiazol-4-yl)-2-(methoxyimino)- acetamido]-3-[(Z)-2-(4-methylthiazol-5-yl)-ethyl]-cephem-4-carboxy lic acid. The susceptibilities of about 1,600 clinical isolates to ME1206 were determined by the agar dilution method. ME1206 showed a broad spectrum of activity against gram-positive and gram-negative bacteria. ME1206 was more active than cefaclor, T-2525, and cefixime against Staphylococcus aureus and Staphylococcus epidermidis. Against gram-negative bacteria, the activity of ME1206 was comparable with that of T-2525, but ME1206 was less active than cefixime. Against Streptococcus pneumoniae, Haemophilus influenzae, and Neisseria gonorrhoeae, ME1206 had high activity (MIC, less than or equal to 0.05 microgram/ml). ME1206 was stable against various beta-lactamases, except beta-lactamases from Providencia rettgeri, Pseudomonas cepacia, and Escherichia coli W3630 (Rms213). The 50% effective doses of ME1207 after oral administration against systemic infections in mice were comparable with those of T-2588 against gram-negative bacteria and about one-fourth that of T-2588 against Staphylococcus aureus Smith.
Sigma-Aldrich
Cefditoren sodium, ≥95% (HPLC)
Sigma-Aldrich
Cefditoren pivoxil, ≥98% (HPLC)