Merck
SUMO co-expression modifies KV 11.1 channel activity.
Acta physiologica (Oxford, England) (2017-09-10)
A B Steffensen, M N Andersen, N Mutsaers, A Mujezinovic, N Schmitt
ABSTRACT
The voltage-gated potassium channel KV 11.1 is the molecular basis for the IKr current, which plays an important role in cardiac physiology. Its malfunction is associated with both inherited and acquired cardiac arrhythmias. Native currents differ from those in experimental models, suggesting additional regulatory mechanisms. We hypothesized that the post-translational modification sumoylation fine-tunes channel activity. The functional effects of sumoylation on KV 11.1 were addressed by employing two-electrode voltage-clamp (TEVC) experiments in Xenopus laevis oocytes. Site-directed mutagenesis enabled a further analysis of the SUMO-target amino acids. We assessed protein expression levels and used confocal imaging for localization studies. Co-expression with Ubc9 and SUMO alters the electrophysiological properties of KV 11.1 leading to a decrease in steady-state current amplitude largely due to faster inactivation and alteration of deactivation kinetics. We identified three lysines (K21, K93 and K116) in the PAS domain as the putative SUMO-targets. This study indicates KV 11.1 as a sumoylation target and offers three main targets: K21, K93, and K116. Furthermore, it proposes an underlying mechanism for the observed kinetic impact of the PAS domain.
Sigma-Aldrich
Anti-UBE2I antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-SUMO-2 antibody produced in rabbit, ~0.6 mg/mL, affinity isolated antibody, buffered aqueous solution