Merck
  • Styrene-Butadiene Rubber by Miniemulsion Polymerization Using In Situ Generated Surfactant.
Styrene-Butadiene Rubber by Miniemulsion Polymerization Using In Situ Generated Surfactant.
Polymers (2020-07-08)
Anderson M S Medeiros, Elodie Bourgeat-Lami, Timothy F L McKenna
ABSTRACT
An alternative approach for the synthesis of styrene butadiene rubber (SBR) copolymer latexes was explored in order to obtain low gel fractions and high solid contents. The ultra-turrax-assisted miniemulsion stabilized by in situ surfactant generation was adopted as the main strategy since this technique can inhibit the eventual presence of secondary nucleation producing polybutadiene particles and also control the cross-linking degree. Styrene monomer was first miniemulsified using an ultra-turrax and in situ generated surfactant using either hexadecane (HD) or octadecyl acrylate (ODA) as the hydrophobe. Dynamic light scattering (DLS) measurements of droplet size indicated faster stabilization and the production of smaller droplet diameters ca. 190 nm (PdI = 0.08) when employing in situ generated potassium oleate (K-Oleate) in comparison to SDS-based miniemulsions. High butadiene-level SBR latexes with ca. 50% solids content, a glass transition temperature (Tg) of -52 °C, and a butadiene to styrene weight ratio of 75:25, were then obtained using the miniemulsion droplets as seeds. Turbiscan and DLS measurements revealed a very stable resulting latex with SBR particle diameter of ca. 220 nm and a low polydispersity index (PdI). Secondary nucleation was prevented as indicated by the low Np/Nd value. Cryo-TEM images showed a narrow distribution of particle size as well as the absence of agglomeration. The gel content was below 10% when tert-dodecyl mercaptan (t-DM) was used as chain transfer agent (CTA).
Sigma-Aldrich
Octadecyl acrylate, contains 200 ppm monomethyl ether hydroquinone as inhibitor, 97%
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
2,2′-Azobis(2-methylbutyronitrile), ≥98.0%