Merck
  • A Toolbox Approach To Construct Broadly Applicable Metal-Free Catalysts for Photoredox Chemistry: Deliberate Tuning of Redox Potentials and Importance of Halogens in Donor-Acceptor Cyanoarenes.
A Toolbox Approach To Construct Broadly Applicable Metal-Free Catalysts for Photoredox Chemistry: Deliberate Tuning of Redox Potentials and Importance of Halogens in Donor-Acceptor Cyanoarenes.
Journal of the American Chemical Society (2018-10-03)
Elisabeth Speckmeier, Tillmann G Fischer, Kirsten Zeitler
ABSTRACT
The targeted choice of specific photocatalysts has been shown to play a critical role for the successful realization of challenging photoredox catalytic transformations. Herein, we demonstrate the successful implementation of a rational design strategy for a series of deliberate structural manipulations of cyanoarene-based, purely organic donor-acceptor photocatalysts, using 1,2,3,5-tetrakis(carbazol-9-yl)-4,6-dicyanobenzene (4CzIPN) as a starting point. Systematic modifications of both the donor substituents as well as the acceptors' molecular core allowed us to identify strongly oxidizing as well as strongly reducing catalysts (e.g., for an unprecedented detriflation of unactivated naphthol triflate), which additionally offer remarkably balanced redox potentials with predictable trends. Especially halogen arene core substitutions are instrumental for our targeted alterations of the catalysts' redox properties. Based on their preeminent electrochemical and photophysical characteristics, all novel, purely organic photoredox catalysts were evaluated in three challenging, mechanistically distinct classes of benchmark reactions (either requiring balanced, highly oxidizing or strongly reducing properties) to demonstrate their enormous potential as customizable photocatalysts, that outperform and complement prevailing typical best photocatalysts.
Sigma-Aldrich
3CzClIPN
Sigma-Aldrich
3DPA2FBN, ≥95%
Sigma-Aldrich
3DPAFIPN, ≥95%