
Triphenyl phosphite

Synonym(s):
(PhO)3P, P(OPh)3, Triphenoxyphosphine, Triphenyl phosphite
Linear Formula:
(C6H5O)3P
CAS Number:
Molecular Weight:
310.28
Products
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing