
TLC Aluminum Sheets

Synonym(s):
HPTLC (High Performance TLC) Aluminum Sheets
Products
Product Category
Brand
Manufacturer
Length (cm)
Quality Segment
Fluorescence
Matrix Material
Support Material
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-7 of 7
Compare
Product Number
Description
Pricing