
Spirotetramat Metabolite BYI08330-mono-hydroxy

Synonym(s):
cis-3-(2,5-Dimethylphenyl)-4-hydroxy-8-methoxy-1-azaspiro[4.5]decan-2-one
Empirical Formula (Hill Notation):
C18H25NO3
CAS Number:
Molecular Weight:
303.40
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique