
Sphingosylphosphorylcholine

Synonym(s):
(2S,3R,4E)-2-Amino-4-octadecene-3-hydroxy-1-phosphocholine, 2-[[[(2-Amino-3-hydroxy-4-octadecenyl)oxy]hydroxy-phosphinyl]oxy]-N,N,N-trimethylethanaminium inner salt, D-erythro-Sphingosine phosphocholine, Lyso SM (d18:1), Lysosphingomyelin, Sphingosylphosphoryl-choline
Empirical Formula (Hill Notation):
C23H49N2O5P
CAS Number:
Molecular Weight:
464.62
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-2 of 2