Skip to Content
Merck
(R)-(+)-Styrene oxide ChiPros®, produced by BASF, ≥98%

(R)-(+)-Styrene oxide

(R)-(+)-Styrene oxide ChiPros®, produced by BASF, ≥98%
Synonym(s):
(R)-(+)-Phenyloxirane, (R)-Phenylethylene oxide
Empirical Formula (Hill Notation):
C8H8O
CAS Number:
Molecular Weight:
120.15

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2