
Phenylbutyrate Related Compound B

Synonym(s):
α-Tetralone, 3,4-Dihydro-1(2H)-naphthalenone
Empirical Formula (Hill Notation):
C10H10O
CAS Number:
Molecular Weight:
146.19
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2