Skip to Content
Merck
Phenylbutazone certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Phenylbutazone

Phenylbutazone certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
4-Butyl-1,2-diphenyl-3,5-pyrazolidinedione, Phenylbutazone
Empirical Formula (Hill Notation):
C19H20N2O2
CAS Number:
Molecular Weight:
308.37
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique