
Nitrofurantoin Related Compound A

Synonym(s):
3-(5-Nitrofurfurylideneamino)hydantoic acid, N-(Aminocarbonyl)-N-{[(5-nitro-2-furanyl)methylene]amino}glycine
Empirical Formula (Hill Notation):
C8H8N4O6
CAS Number:
Molecular Weight:
256.17
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing