![(−)-Nicotine solution 1.0 mg/mL, analytical standard, for drug analysis](/deepweb/assets/sigmaaldrich/product/structures/184/935/71e5d704-3ebe-4873-88dd-76444d306315/640/71e5d704-3ebe-4873-88dd-76444d306315.png)
(−)-Nicotine solution
![(−)-Nicotine solution 1.0 mg/mL, analytical standard, for drug analysis](/deepweb/assets/sigmaaldrich/product/structures/184/935/71e5d704-3ebe-4873-88dd-76444d306315/640/71e5d704-3ebe-4873-88dd-76444d306315.png)
Empirical Formula (Hill Notation):
C10H14N2
CAS Number:
Molecular Weight:
162.23
Products
Product Category
Brand
Formula Weight
Gene Alias
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing