
Methyl linoleate

Synonym(s):
Linoleic acid methyl ester, Methyl cis,cis-9,12-octadecadienoate, Methyl linoleate
Linear Formula:
CH3(CH2)3(CH2CH=CH)2(CH2)7CO2CH3
CAS Number:
Molecular Weight:
294.47
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Functional Group
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique