Skip to Content
Merck
(−)-Menthol primary reference standard

Menthol

(−)-Menthol primary reference standard
Synonym(s):
(−)-Menthol, (1R,2S,5R)-2-Isopropyl-5-methylcyclohexanol, 5-Methyl-2-(1-methylethyl)cyclohexanol
Empirical Formula (Hill Notation):
C10H20O
CAS Number:
Molecular Weight:
156.27
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique