
Luteolin 7-glucuronide

Synonym(s):
Luteolin-7-O-β-D-glucuronide, 2-(3,4-Dihydroxyphenyl)-5-hydroxy-4-oxo-4H-1-benzopyran-7-yl-β-D-glucopyranosiduronic acid, 3′,4′,5,7-Tetrahydroxy-7-β-D-glucopyranuronoside flavone, 3′,4′,5,7-Tetrahydroxyflavone 7-glucuronide, Cyanidenon 7-glucuronide, Cyanidenon-7-O-β-D-glucuronic acid, Luteolin 7-O-β-D-glucuronopyranoside
Empirical Formula (Hill Notation):
C21H18O12
CAS Number:
Molecular Weight:
462.36
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing