
Lovastatin Related Compound A

Synonym(s):
Dihydrolovastatin
Empirical Formula (Hill Notation):
C24H38O5
CAS Number:
Molecular Weight:
406.56
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2