
Loratadine Related Compound C

Synonym(s):
8-Chloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-one
Empirical Formula (Hill Notation):
C14H10ClNO
CAS Number:
Molecular Weight:
243.69
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2