
L-(−)-Mannose

Synonym(s):
L-Mannopyranose
Empirical Formula (Hill Notation):
C6H12O6
Products
Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Optical Isomer
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing