Skip to Content
Merck
L-Lysine-2-15N hydrochloride endotoxin tested, 98 atom % 15N

L-Lysine-2-15N hydrochloride

L-Lysine-2-15N hydrochloride endotoxin tested, 98 atom % 15N
Linear Formula:
H2N(CH2)4CH(15NH2)CO2H·HCl
CAS Number:
Molecular Weight:
183.64

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Isotope
Markush Class
Markush Group
Optical Isomer
Technique
Showing 1-2 of 2