
Levonorgestrel

Synonym(s):
D(−)-Norgestrel, 13β-Ethyl-17α-ethynyl-17β-hydroxygon-4-en-3-one, 17α-Ethynyl-18-homo-19-nor-testosterone, 18,19-Dinor-13β-ethyl-17β-hydroxy-4-pregnen-20-yn-3-one, 18,19-Dinor-4-pregnen-20-yn-3-one, Levonorgestrel, (17α)-13-Ethyl-17-hydroxy-18,19-dinorpregn-4-en-20-yn-3-one, 13-Ethyl-17-ethynyl-17β-hydroxy-4-gonen-3-one, 13-Ethyl-17-hydroxy-18,19-dinor-17α-pregn-4-en-20-yn-3-one, D(−)-Norgestrel, D-Norgestrel, LNG
Empirical Formula (Hill Notation):
C21H28O2
Products
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing