Skip to Content
Merck
(−)-Isoledene ≥95.0% (sum of enantiomers, GC)

(−)-Isoledene

(−)-Isoledene ≥95.0% (sum of enantiomers, GC)
Empirical Formula (Hill Notation):
C15H24
CAS Number:
Molecular Weight:
204.35

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing