
Isoimperatorin

Synonym(s):
4-(3-Methylbut-2-enyloxy)furo[3,2-g]chromen-7-one, 4-[(3-Methyl-2-buten-1-yl)oxy]-7H-furo[3,2-g][1]benzopyran-7-one
Empirical Formula (Hill Notation):
C16H14O4
CAS Number:
Molecular Weight:
270.28
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing