Glutamine
Synonym(s):
L-Glutamine, (S)-2,5-Diamino-5-oxopentanoic acid, L-Glutamic acid 5-amide, Levoglutamide
Linear Formula:
H2NCOCH2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
146.14
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing