
Gamma Cyclodextrin

Synonym(s):
γ-Cyclodextrin, Cyclomaltooctaose, Cyclooctaamylose, Schardinger γ-Dextrin
Empirical Formula (Hill Notation):
C48H80O40
CAS Number:
Molecular Weight:
1297.12
Products
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing