
Famotidine Related Compound E

Synonym(s):
N,N′′′-[Dithiobis(methylene-4,2-thiazolediyl)]bisguanidine, 2,2′-[4,4′-Disulfanediylbis(methylene)bis(thiazole-4,2-diyl)]diguanidine
Empirical Formula (Hill Notation):
C10H14N8S4
CAS Number:
Molecular Weight:
374.53
Products
Brand
Biological Source
Color
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2