
Ethylhexadecyldimethylammonium bromide

Synonym(s):
Cetyldimethylethylammonium bromide, Dimethylethylhexadecylammonium bromide, Ethylhexadecyldimethylammonium bromide
Linear Formula:
C2H5N(CH3)2C16H33Br
CAS Number:
Molecular Weight:
378.47
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Chemical Composition
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing