
Entacapone impurity A

Synonym(s):
(Z)-Entacapone, (2Z)-2-Cyano-3-(3,4-dihydroxy-5-nitrophenyl)-N,N-diethyl-2-propenamide
Empirical Formula (Hill Notation):
C14H15N3O5
CAS Number:
Molecular Weight:
305.29
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing