
Endfitting with fixed 10 μm frit for all 7.5 mm I.D. TSKgel® stainless steel columns

Synonym(s):
TSKgel® Hardware and Accessories
Products
Manufacturer
Product Line
Package Size
Pore size (µm)
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing