
Econazole Related Compound A

Synonym(s):
(1RS)-1-(2,4-Dichlorophenyl)-2-(-1H-imidazol-1-yl)ethanol
Empirical Formula (Hill Notation):
C11H10Cl2N2O
CAS Number:
Molecular Weight:
257.12
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2