
(±)-Bay K8644

Synonym(s):
1,4-Dihydro-2,6-dimethyl-5-nitro-4-(2-[trifluoromethyl]phenyl)pyridine-3-carboxylic acid methyl ester
Empirical Formula (Hill Notation):
C16H15F3N2O4
CAS Number:
Molecular Weight:
356.30
Products
Brand
Color
Formula Weight
Gene Alias
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing