D-(−)-3-Phosphoglyceric acid disodium salt
Synonym(s):
(−)-Disodium D-3-phosphoglycerate, D-Glycerate 3-phosphate disodium salt
Empirical Formula (Hill Notation):
C3H5Na2O7P
CAS Number:
Molecular Weight:
230.02
Products
Product Category
Brand
Sustainability
Biological Source
Color
Feature
Formula Weight
Physical Form
Purity
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing