Skip to Content
Merck
Benazepril Related Compound A Pharmaceutical Secondary Standard; Certified Reference Material

Benazepril Related Compound A

Benazepril Related Compound A Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
(3R)-3-[[(1R)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]-2,3,4,5-tetrahydro-2-oxo-1H-1-benzazepine-1-acetic acid hydrochloride
Empirical Formula (Hill Notation):
C24H28N2O5 · HCl
CAS Number:
Molecular Weight:
460.95

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade