
5-Hydroxythiabendazole

Synonym(s):
2-(4-Thiazolyl)-5-benzimidazolol, 5-Hydroxy-2-(4-thiazolyl)benzimidazole
Empirical Formula (Hill Notation):
C10H7N3OS
CAS Number:
Molecular Weight:
217.25
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing