
4-Bromo-calcium Ionophore A23187

Synonym(s):
4-Bromo-A23187, 4-Bromo-calcimycin
Empirical Formula (Hill Notation):
C29H36BrN3O6
CAS Number:
Molecular Weight:
602.52
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing