
1-Amino-2-methylpropan-2-ol

Empirical Formula (Hill Notation):
C4H11NO
CAS Number:
Molecular Weight:
89.14
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing