Skip to Content
Merck
All Photos(1)

Documents

HUMANNCD8-0002090

Sigma-Aldrich

Frozen Human Normal CD8+/CD45RA+ Naïve Cytotoxic T Cells

human peripheral blood (CD8+/CD45RA+ Naïve Cytotoxic T Cells)

Synonym(s):

CD8+/CD45RA+ Cells, Immune Cells, Naïve Cytotoxic T Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

Frozen Human Normal CD8+/CD45RA+ Naïve Cytotoxic T Cells, 5 Million Cells/per vial

biological source

human peripheral blood (CD8+/CD45RA+ Naïve Cytotoxic T Cells)

Quality Level

form

frozen liquid

packaging

vial of 5 Million cells

manufacturer/tradename

BioIVT

technique(s)

cell culture | mammalian: suitable

color

cloudy

storage temp.

-140 to -196°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.

We are pleased to offer these products from BioIVT.

Naive T cells express the highest molecular weight isoform of CD45, CD45 RA. Those which express CD8 are destined to become cytotoxic T cells. Naive Cytotoxic T cells have not yet been exposed to antigens

Human Peripheral Blood CD8+/CD45RA+ Naive Cytotoxic T cells are isolated from peripheral blood mononuclear cells by negative immunomagnetic selection. All peripheral blood is collected from fully consented IRB approved donors.

Packaging

5 Million Cells/per vial

Other Notes

Thaw before use.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service