Skip to Content
Merck
All Photos(1)

Key Documents

C7415

Sigma-Aldrich

Anti-Cat Serum antibody produced in rabbit

whole antiserum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

secondary antibodies

clone

polyclonal

contains

15 mM sodium azide

species reactivity

cat

technique(s)

immunoelectrophoresis: suitable

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Cat serum protein comprises immunoglobulins (Ig) including IgA, IgM, and IgG. IgG is majorly secreted in precolostral sera and cerebrospinal fluid. IgA and IgM is present in bile, nasal secretions, colostral whey and postcolostral sera.
Pooled normal cat serum was used as an immunogen in rabbit to develop anti-cat whole serum antibody.

Specificity

Strong reactivity with normal cat serum has been deter-mined by immunoelectrophoresis (IEP). This antiserum has not been assayed for interspecies cross reactivity.

Application

Anti-Cat Serum antibody produced in rabbit was fluoresceinated and used in indirect immunofluorescence assay for the detection of feline immunodeficiency virus.

Physical form

Rabbit anti-Cat Serum is provided as a liquid containing 0.1% sodium azide (see MSDS) as preservative.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, the solution may be frozen in working aliquots. Repeated freezing and thawing is not recommended. Storage in "frost-free" freezers not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Feline immunoglobulins
Schultz RD, et al.
Infection and Immunity, 9(2), 391-393 (1974)
A Poli et al.
Journal of clinical microbiology, 30(8), 2038-2041 (1992-08-01)
The saliva of cats infected with feline immunodeficiency virus was examined for total immunoglobulin content and antiviral antibodies. Seropositive cats showed an increase in salivary immunoglobulin G levels, which was only partly attributable to the enhanced prevalence of oral inflammatory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service