RCHFA4VP1
RCS® High Flow Touch Validation Protocol
A4 Format, Microbial Air Sampler Validation Guide
Synonym(s):
Air Sampler Validation Protocol, Microbial Air Sampler Validation Protocol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Quality Level
manufacturer/tradename
RCS®
input
sample type pharmaceutical(s)
application(s)
industrial qc
compatibility
for use with RCS® High Flow Touch Microbial Air Samplers.
shipped in
ambient
General description
RCS® High Flow Touch Validation protocol is a ready to use guide designed for qualifying the RCS® High Flow Touch Microbial Air Samplers before moving to routine testing It covers the installation qualification, operational qualification and performance qualification. Air monitoring is a critical control step laid down by the regulatory bodies for ensuring product quality inside cleanrooms.
The validation protocol in A4 format ensures regulatory compliance of the air samplers and guarantees consistent performance.
The validation protocol in A4 format ensures regulatory compliance of the air samplers and guarantees consistent performance.
Application
RCS® High Flow Touch Validation protocol is used to validate RCS® High Flow Touch Microbial Air Samplers for consistent performance and regulatory compliance.
Legal Information
RCS is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service