Skip to Content
Merck
All Photos(1)

Key Documents

8.57001

Sigma-Aldrich

Activator Reagent for DNA Synthesis

0.3M BMT in MeCN, Novabiochem®

Synonym(s):

0.3M BMT in MeCN, Novabiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116006
NACRES:
NA.22

Quality Level

product line

Novabiochem®

reaction suitability

reaction type: Coupling Reactions
reagent type: catalyst

General description

Pre-mixed ready-to-use Activator Reagent
  • 857001.1000: Compatible with Cytiva ÄKTA DNA Synthesisers. Stand alone from Instrument (GL-45 Neck Finish)
  • 857001.2500: Compatible with Cytiva ÄKTA DNA Synthesisers. Stand alone from Instrument (GL-45 Neck Finish)
  • 857001.4000: Compatible with MerMade DNA Synthesisers. Stand alone from Instrument (38mm Neck Finish)

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

35.6 °F - (Acetonitrile)

Flash Point(C)

2 °C - (Acetonitrile)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service