Skip to Content
Merck
All Photos(2)

Key Documents

804312

Sigma-Aldrich

2,2-difluoroethylamine hydrochloride

Synonym(s):

2,2-difluoro-1-ethanamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H6ClF2N
CAS Number:
Molecular Weight:
117.53
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

Quality Level

form

powder

mp

132-137 °C (w/decomp)

storage temp.

2-8°C

SMILES string

Cl.NCC(F)F

InChI

1S/C2H5F2N.ClH/c3-2(4)1-5;/h2H,1,5H2;1H

InChI key

YSVKPVHTHFECBE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,2-Difluoroethylamine hydrochloride was recently reported to be used as a precursor for the "elusive" difluoromethyl diazomethane species (CF2HCHN2). The difluoromethyl diazomethane intermediate can be generated in situ in the presence of tert-butyl nitrite and a catalytic amount of acetic acid.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service