Merck
  • Selectivity assessment of popular stationary phases for open-tubular column gas chromatography.
Selectivity assessment of popular stationary phases for open-tubular column gas chromatography.
Journal of chromatography. A (2001-04-20)
C F Poole, Q Li, W Kiridena, W W Koziol
ABSTRACT
The solvation parameter model is used to study the influence of temperature and composition on the selectivity of nine poly(siloxane) and two poly(ethylene glycol) stationary phase chemistries for open-tubular column gas chromatography. A database of system constants for the temperature range 60-140 degrees C was constructed from literature values with additional results determined for HP-50+, DB-210, DB-1701, DB-225 and SP-2340 columns. The general contribution of monomer composition (methyl, phenyl, cyanopropyl, and trifluoropropyl substituents) on the capacity of poly(siloxane) stationary phases for dispersion, electron lone pair, dipole-type and hydrogen-bond interactions is described. The selectivity coverage of the open-tubular column stationary phases is compared with a larger database for packed column stationary phases at a reference temperature of 120 degrees C. The open-tubular column stationary phases provide reasonable coverage of the range of dipole-type and hydrogen-bond base interactions for non-ionic packed column stationary phases. Deficiencies are noted in the coverage of electron lone pair interactions. None of the open-tubular column stationary phases are hydrogen-bond acids. The system constants are shown to change approximately linearly with temperature over the range 60-140 degrees C. The intercepts and slopes of these plots are used to discuss the influence of temperature on stationary phase selectivity.
Product Description
Supelco
SP®-2340 Capillary GC Column, L × I.D. 60 m × 0.32 mm, df 0.20 μm
Supelco
SP®-2340 Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 0.20 μm
Supelco
SP®-2340 Capillary GC Column, L × I.D. 60 m × 0.25 mm, df 0.20 μm