Merck
  • Efficient modification of λ-DNA substrates for single-molecule studies.
Efficient modification of λ-DNA substrates for single-molecule studies.
Scientific reports (2017-05-20)
Yoori Kim, Armando de la Torre, Andrew A Leal, Ilya J Finkelstein
ABSTRACT
Single-molecule studies of protein-nucleic acid interactions frequently require site-specific modification of long DNA substrates. The bacteriophage λ is a convenient source of high quality long (48.5 kb) DNA. However, introducing specific sequences, tertiary structures, and chemical modifications into λ-DNA remains technically challenging. Most current approaches rely on multi-step ligations with low yields and incomplete products. Here, we describe a molecular toolkit for rapid preparation of modified λ-DNA. A set of PCR cassettes facilitates the introduction of recombinant DNA sequences into the λ-phage genome with 90-100% yield. Extrahelical structures and chemical modifications can be inserted at user-defined sites via an improved nicking enzyme-based strategy. As a proof-of-principle, we explore the interactions of S. cerevisiae Proliferating Cell Nuclear Antigen (yPCNA) with modified DNA sequences and structures incorporated within λ-DNA. Our results demonstrate that S. cerevisiae Replication Factor C (yRFC) can load yPCNA onto 5'-ssDNA flaps, (CAG)
Sigma-Aldrich
Monoclonal ANTI-FLAG® BioM2 antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous glycerol solution