Merck
  • Proteome analysis of multidrug-resistant, breast cancer-derived microparticles.
Proteome analysis of multidrug-resistant, breast cancer-derived microparticles.
Journal of extracellular vesicles (2014-09-11)
Deep Pokharel, Matthew P Padula, Jamie F Lu, Jessica L Tacchi, Frederick Luk, Steven P Djordjevic, Mary Bebawy
ABSTRACT
Cancer multidrug resistance (MDR) occurs when cancer cells evade the cytotoxic actions of chemotherapeutics through the active efflux of drugs from within the cells. Our group have previously demonstrated that multidrug-resistant breast cancer cells spontaneously shed microparticles (MPs) and that these MPs can transfer resistance to drug-responsive cells and confer MDR on those cells in as little as 4 h. Furthermore, we also showed that, unlike MPs derived from leukaemia cells, breast cancer-derived MPs display a tissue selectivity in the transfer of P-glycoprotein (P-gp), transferring the resistance protein only to malignant breast cells. This study aims to define the proteome of breast cancer-derived MPs in order to understand the differences in protein profiles between those shed from drug-resistant versus drug-sensitive breast cancer cells. In doing so, we detail the protein cargo required for the intercellular transfer of MDR to drug-sensitive recipient cells and the factors governing the transfer selectivity to malignant breast cells. We describe the first proteomic analysis of MPs derived from human breast cancer cells using SDS PAGE and liquid chromatography-tandem mass spectrometry (LC/MS/MS), in which we identify 120 unique proteins found only in drug-resistant, breast cancer-derived MPs. Our results demonstrate that the MP-mediated transfer of P-gp to recipient cells occurs alongside CD44; the Ezrin, Radixin and Moesin protein family (ERM); and cytoskeleton motor proteins within the MP cargo.
Sigma-Aldrich
Monoclonal Anti-Moesin antibody produced in mouse, clone 38/87, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Monoclonal Anti-P-Glycoprotein (MDR) antibody produced in mouse, clone F4, ascites fluid