Merck
  • Molecular mechanism of apolipoprotein E binding to lipoprotein particles.
Molecular mechanism of apolipoprotein E binding to lipoprotein particles.
Biochemistry (2009-02-13)
David Nguyen, Padmaja Dhanasekaran, Michael C Phillips, Sissel Lund-Katz
ABSTRACT
The exchangeability of apolipoprotein (apo) E between lipoprotein particles such as very low-density lipoprotein (VLDL) and high-density lipoprotein (HDL) is critical for lipoprotein metabolism, but despite its importance, the kinetics and mechanism of apoE-lipoprotein interaction are not known. We have used surface plasmon resonance (SPR) to monitor in real time the reversible binding of apoE to human VLDL and HDL(3); biotinylated lipoproteins were immobilized on a streptavidin-coated SPR sensor chip, and solutions containing various human apoE molecules at different concentrations were passed across the surface. Analysis of the resultant sensorgrams indicated that the apoE3-lipoprotein interaction is a two-step process. After an initial interaction, the second slower step involves opening of the N-terminal helix bundle domain of the apoE molecule. Destabilization of this domain leads to more rapid interfacial rearrangement which is seen when the lipoprotein binding of apoE4 is compared to that of apoE3. The resultant differences in interfacial packing seem to underlie the differing abilities of apoE4 and apoE3 to bind to VLDL and HDL(3). The measured dissociation constants for apoE binding to these lipoprotein particles are in the micromolar range. C-Terminal truncations of apoE to remove the lipid binding region spanning residues 250-299 reduce the level of binding to both types of lipoprotein, but the effect is weaker with HDL(3); this suggests that protein-protein interactions are important for apoE binding to this lipoprotein while apoE-lipid interactions are more significant for VLDL binding. The two-step mechanism of lipoprotein binding exhibited by apoE is likely to apply to other members of the exchangeable apolipoprotein family.
Sigma-Aldrich
Anti-APOE (C-terminal) antibody produced in rabbit
Pricing and availability is not currently available.