Merck
  • Plasticity of calcium signaling cascades in human embryonic stem cell-derived neural precursors.
Plasticity of calcium signaling cascades in human embryonic stem cell-derived neural precursors.
Stem cells and development (2013-01-09)
Oksana Forostyak, Nataliya Romanyuk, Alexei Verkhratsky, Eva Sykova, Govindan Dayanithi
ABSTRACT
Human embryonic stem cell-derived neural precursors (hESC NPs) are considered to be a promising tool for cell-based therapy in central nervous system injuries and neurodegenerative diseases. The Ca(2+) ion is an important intracellular messenger essential for the regulation of various cellular functions. We investigated the role and physiology of Ca(2+) signaling to characterize the functional properties of CCTL14 hESC NPs during long-term maintenance in culture (in vitro). We analyzed changes in cytoplasmic Ca(2+) concentration ([Ca(2+)]i) evoked by high K(+), adenosine-5'-triphosphate (ATP), glutamate, γ-aminobutyric acid (GABA), and caffeine in correlation with the expression of various neuronal markers in different passages (P6 through P10) during the course of hESC differentiation. We found that only differentiated NPs from P7 exhibited significant and specific [Ca(2+)]i responses to various stimuli. About 31% of neuronal-like P7 NPs exhibited spontaneous [Ca(2+)]i oscillations. Pharmacological and immunocytochemical assays revealed that P7 NPs express L- and P/Q-type Ca(2+) channels, P2X2, P2X3, P2X7, and P2Y purinoreceptors, glutamate receptors, and ryanodine (RyR1 and RyR3) receptors. The ATP- and glutamate-induced [Ca(2+)]i responses were concentration-dependent. Higher glutamate concentrations (over 100 μM) caused cell death. Responses to ATP were observed in the presence or in the absence of extracellular Ca(2+). These results emphasize the notion that with time in culture, these cells attain a transient period of operative Ca(2+) signaling that is predictive of their ability to act as stem elements.
Sigma-Aldrich
Bovine Serum Albumin, fatty acid free, low endotoxin, lyophilized powder, BioReagent, suitable for cell culture, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
γ-Aminobutyric acid, BioXtra, ≥99%
Sigma-Aldrich
Nicardipine hydrochloride, powder, ≥98%
Sigma-Aldrich
Laminin from human fibroblasts, cell culture derived, liquid, sterile-filtered
Sigma-Aldrich
Accutase® solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Nickel(II) chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Cadmium chloride, 99.99% trace metals basis
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, BioXtra, ≥99% (HPLC), from microbial
Sigma-Aldrich
Kainic acid monohydrate, ≥99% (TLC)
Sigma-Aldrich
α,β-Methyleneadenosine 5′-triphosphate lithium salt, ≥93% (HPLC), solid
Sigma-Aldrich
Adenosine, suitable for cell culture, BioReagent
Sigma-Aldrich
L-Glutamic acid potassium salt monohydrate, BioReagent, suitable for insect cell culture, ≥99% (HPLC)
Sigma-Aldrich
Pyridoxal phosphate-6-azo(benzene-2,4-disulfonic acid) tetrasodium salt hydrate, solid, ≥98% (HPLC)
Sigma-Aldrich
Suramin sodium salt, ≥98% (TLC)
Sigma-Aldrich
Anti-Glutamine Synthetase Antibody, clone GS-6, clone GS-6, Chemicon®, from mouse